Assay Details
| Gene Symbol: bam-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026638.4 | NP_001021809.1 | 12 - 13 | 2924 | 93 | |
| GenBank mRNA | AY373340.1 | - | 12 - 13 | 2890 | 93 | |
More Information
Set Membership: |
|

bam-2
Genomic Map