Assay Details
| Gene Symbol: Y54E10BR.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058689.4 | NP_491090.1 | 1 - 2 | 78 | 89 | |
| Gene Symbol: Y54E10BR.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058690.5 | NP_491091.1 | 1 - 2 | 88 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_058690.5 transcripts.
Set Membership: |
|

Y54E10BR.4
Genomic Map