Assay Details
| Gene Symbol: ztf-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058695.7 | NP_491096.1 | 9 - 10 | 1174 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ztf-23| Gene Symbol: ztf-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058695.7 | NP_491096.1 | 9 - 10 | 1174 | 105 | |
Set Membership: |
|