Assay Details
| Gene Symbol: pign-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001262367.2 | NP_001249296.1 | 3 - 4 | 626 | 83 | |
| NM_001262368.4 | NP_001249297.1 | 3 - 4 | 2159 | 83 | ||
More Information
Set Membership: |
|

pign-1
Genomic Map