Assay Details
| Gene Symbol: pign-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001262367.2 | NP_001249296.1 | 4 - 5 | 779 | 82 | |
| NM_001262368.4 | NP_001249297.1 | 4 - 5 | 2312 | 82 | ||
More Information
Set Membership: |
|

pign-1
Genomic Map