Assay Details
| Gene Symbol: zip-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047238.7 | NP_001040703.1 | 1 - 2 | 702 | 73 | |
| NM_001381600.2 | NP_001368002.1 | 3 - 4 | 1063 | 73 | ||
More Information
Set Membership: |
|

zip-4
Genomic Map