Assay Details
| Gene Symbol: ace-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058740.6 | NP_491141.1 | 4 - 5 | 807 | 64 | |
| GenBank mRNA | AF025378.3 | - | 3 - 4 | 541 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ace-2| Gene Symbol: ace-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058740.6 | NP_491141.1 | 4 - 5 | 807 | 64 | |
| GenBank mRNA | AF025378.3 | - | 3 - 4 | 541 | 64 | |
Set Membership: |
|