Assay Details
| Gene Symbol: ace-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058740.6 | NP_491141.1 | 8 - 9 | 1866 | 116 | |
| GenBank mRNA | AF025378.3 | - | 7 - 8 | 1600 | 116 | |
More Information
Set Membership: |
|

ace-2
Genomic Map