Assay Details
| Gene Symbol: cdap-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058742.6 | NP_491143.3 | 4 - 5 | 729 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdap-2| Gene Symbol: cdap-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058742.6 | NP_491143.3 | 4 - 5 | 729 | 85 | |
Set Membership: |
|