Assay Details
| Gene Symbol: cdk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026366.3 | NP_001021537.2 | 6 - 7 | 988 | 84 | |
| NM_001350362.3 | NP_001337286.1 | 6 - 7 | 1005 | 84 | ||
More Information
Set Membership: |
|

cdk-2
Genomic Map