Assay Details
| Gene Symbol: crn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058767.8 | NP_491168.1 | 1 - 2 | 127 | 81 | |
| GenBank mRNA | AY303575.1 | - | 1 - 2 | 94 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

crn-1| Gene Symbol: crn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058767.8 | NP_491168.1 | 1 - 2 | 127 | 81 | |
| GenBank mRNA | AY303575.1 | - | 1 - 2 | 94 | 81 | |
Set Membership: |
|