Assay Details
| Gene Symbol: crn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058767.8 | NP_491168.1 | 3 - 4 | 989 | 65 | |
| GenBank mRNA | AY303575.1 | - | 3 - 4 | 956 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

crn-1| Gene Symbol: crn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058767.8 | NP_491168.1 | 3 - 4 | 989 | 65 | |
| GenBank mRNA | AY303575.1 | - | 3 - 4 | 956 | 65 | |
Set Membership: |
|