Assay Details
| Gene Symbol: acd-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058795.4 | NP_491196.3 | 9 - 10 | 1563 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acd-5| Gene Symbol: acd-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058795.4 | NP_491196.3 | 9 - 10 | 1563 | 75 | |
Set Membership: |
|