Assay Details
| Gene Symbol: hus-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058802.7 | NP_491203.1 | 2 - 3 | 124 | 73 | |
| GenBank mRNA | AF117337.1 | - | 2 - 3 | 114 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hus-1| Gene Symbol: hus-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058802.7 | NP_491203.1 | 2 - 3 | 124 | 73 | |
| GenBank mRNA | AF117337.1 | - | 2 - 3 | 114 | 73 | |
Set Membership: |
|