Assay Details
| Gene Symbol: hus-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058802.7 | NP_491203.1 | 5 - 6 | 750 | 157 | |
| GenBank mRNA | AF117337.1 | - | 5 - 6 | 740 | 157 | |
More Information
Set Membership: |
|

hus-1
Genomic Map