Assay Details
| Gene Symbol: larp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058808.5 | NP_491209.3 | 1 - 2 | 233 | 80 | |
| NM_182024.8 | NP_871824.3 | 1 - 2 | 227 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_182024.8 transcripts.
Set Membership: |
|

larp-5
Genomic Map