Assay Details
| Gene Symbol: larp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058808.5 | NP_491209.3 | 3 - 4 | 609 | 102 | |
| NM_182024.8 | NP_871824.3 | 3 - 4 | 603 | 102 | ||
More Information
Set Membership: |
|

larp-5
Genomic Map