Assay Details
| Gene Symbol: larp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058808.5 | NP_491209.3 | 4 - 5 | 742 | 85 | |
| NM_182024.8 | NP_871824.3 | 4 - 5 | 736 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

larp-5| Gene Symbol: larp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058808.5 | NP_491209.3 | 4 - 5 | 742 | 85 | |
| NM_182024.8 | NP_871824.3 | 4 - 5 | 736 | 85 | ||
Set Membership: |
|