Assay Details
| Gene Symbol: asic-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058813.5 | NP_491214.3 | 6 - 7 | 1020 | 75 | |
| Gene Symbol: C32E8.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384008.1 | 6 - 7 | 1028 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384008.1 transcripts.
Set Membership: |
|

asic-1
Genomic Map