Assay Details
| Gene Symbol: guk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129039.6 | NP_001122511.1 | 2 - 3 | 248 | 67 | |
| NM_058842.6 | NP_491243.1 | 2 - 3 | 353 | 67 | ||
More Information
Set Membership: |
|

guk-1
Genomic Map