Assay Details
| Gene Symbol: lrp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058869.5 | NP_491270.2 | 15 - 16 | 12980 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lrp-2| Gene Symbol: lrp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058869.5 | NP_491270.2 | 15 - 16 | 12980 | 74 | |
Set Membership: |
|