Assay Details
| Gene Symbol: unc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026325.2 | NP_001021496.1 | 10 - 11 | 2195 | 89 | |
| NM_001026326.4 | NP_001021497.1 | 10 - 11 | 2083 | 89 | ||
| NM_001381834.2 | NP_001367474.1 | 10 - 11 | 2083 | 89 | ||
| GenBank mRNA | AF048834.1 | - | 10 - 11 | 2195 | 89 | |
| AF048835.1 | - | 10 - 11 | 2195 | 89 | ||
More Information
Set Membership: |
|

unc-73
Genomic Map