Assay Details
| Gene Symbol: unc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026326.4 | NP_001021497.1 | 19 - 20 | 4901 | 58 | |
| GenBank mRNA | AF048835.1 | - | 19 - 20 | 5013 | 58 | |
More Information
Set Membership: |
|

unc-73
Genomic Map