Assay Details
| Gene Symbol: unc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026325.2 | NP_001021496.1 | 20 - 21 | 5098 | 59 | |
| NM_001026327.4 | NP_001021498.1 | 4 - 5 | 632 | 59 | ||
| NM_001026329.6 | NP_001021500.1 | 2 - 3 | 298 | 59 | ||
| NM_001262583.3 | NP_001249512.1 | 2 - 3 | 291 | 59 | ||
| NM_001262584.2 | NP_001249513.1 | 4 - 5 | 679 | 59 | ||
| NM_001375256.3 | NP_001361940.1 | 4 - 5 | 426 | 59 | ||
| NM_001381834.2 | NP_001367474.1 | 20 - 21 | 4986 | 59 | ||
| GenBank mRNA | AF048834.1 | - | 20 - 21 | 5098 | 59 | |
More Information
Set Membership: |
|

unc-73
Genomic Map