Assay Details
| Gene Symbol: unc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026325.2 | NP_001021496.1 | 26 - 27 | 6769 | 93 | |
| NM_001026327.4 | NP_001021498.1 | 10 - 11 | 2303 | 93 | ||
| NM_001262583.3 | NP_001249512.1 | 8 - 9 | 1962 | 93 | ||
| NM_001375256.3 | NP_001361940.1 | 10 - 11 | 2097 | 93 | ||
| GenBank mRNA | AF048834.1 | - | 26 - 27 | 6769 | 93 | |
More Information
Set Membership: |
|

unc-73
Genomic Map