Assay Details
| Gene Symbol: unc-89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025813.2 | NP_001020984.1 | 26 - 27 | 17673 | 63 | |
| NM_001025814.4 | NP_001020985.1 | 26 - 27 | 17482 | 63 | ||
| NM_001025817.4 | NP_001020988.1 | 24 - 25 | 15562 | 63 | ||
| NM_001025818.3 | NP_001020989.1 | 24 - 25 | 15562 | 63 | ||
| NM_001356807.3 | NP_001343717.1 | 25 | 15322 | 63 | ||
| NM_001356808.3 | NP_001343716.1 | 26 | 15763 | 63 | ||
| NM_001356809.3 | NP_001343714.1 | 25 | 17416 | 63 | ||
| NM_001356810.3 | NP_001343718.1 | 25 | 15322 | 63 | ||
| NM_001356811.3 | NP_001343715.1 | 25 | 17416 | 63 | ||
| NM_001356812.3 | NP_001343617.1 | 17 - 18 | 16063 | 63 | ||
| NM_001356813.3 | NP_001343719.1 | 16 - 17 | 13903 | 63 | ||
| NM_001356814.3 | NP_001343711.1 | 17 - 18 | 14344 | 63 | ||
| NM_001356815.3 | NP_001343712.1 | 16 - 17 | 15997 | 63 | ||
| NM_001356816.3 | NP_001343713.1 | 15 - 16 | 14143 | 63 | ||
More Information
Set Membership: |
|

unc-89
Genomic Map