Assay Details
| Gene Symbol: unc-89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025813.2 | NP_001020984.1 | 29 - 30 | 18339 | 62 | |
| NM_001025814.4 | NP_001020985.1 | 29 - 30 | 18148 | 62 | ||
| NM_001025817.4 | NP_001020988.1 | 27 - 28 | 16228 | 62 | ||
| NM_001025818.3 | NP_001020989.1 | 27 - 28 | 16228 | 62 | ||
| NM_001356807.3 | NP_001343717.1 | 27 | 15988 | 62 | ||
| NM_001356808.3 | NP_001343716.1 | 28 | 16429 | 62 | ||
| NM_001356809.3 | NP_001343714.1 | 27 | 18082 | 62 | ||
| NM_001356810.3 | NP_001343718.1 | 27 | 15988 | 62 | ||
| NM_001356811.3 | NP_001343715.1 | 27 | 18082 | 62 | ||
| NM_001356812.3 | NP_001343617.1 | 20 - 21 | 16729 | 62 | ||
| NM_001356813.3 | NP_001343719.1 | 19 - 20 | 14569 | 62 | ||
| NM_001356814.3 | NP_001343711.1 | 20 - 21 | 15010 | 62 | ||
| NM_001356815.3 | NP_001343712.1 | 19 - 20 | 16663 | 62 | ||
| NM_001356816.3 | NP_001343713.1 | 18 - 19 | 14809 | 62 | ||
More Information
Set Membership: |
|

unc-89
Genomic Map