Assay Details
| Gene Symbol: unc-89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025813.2 | NP_001020984.1 | 6 - 7 | 1166 | 84 | |
| NM_001025814.4 | NP_001020985.1 | 6 - 7 | 975 | 84 | ||
| NM_001025817.4 | NP_001020988.1 | 6 - 7 | 975 | 84 | ||
| NM_001025818.3 | NP_001020989.1 | 6 - 7 | 975 | 84 | ||
| NM_001356807.3 | NP_001343717.1 | 5 | 975 | 84 | ||
| NM_001356808.3 | NP_001343716.1 | 5 | 975 | 84 | ||
| NM_001356809.3 | NP_001343714.1 | 5 | 975 | 84 | ||
| NM_001356810.3 | NP_001343718.1 | 5 | 975 | 84 | ||
| NM_001356811.3 | NP_001343715.1 | 5 | 975 | 84 | ||
More Information
Set Membership: |
|

unc-89
Genomic Map