Assay Details
| Gene Symbol: unc-89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025814.4 | NP_001020985.1 | 45 - 46 | 23691 | 94 | |
| NM_001025818.3 | NP_001020989.1 | 43 - 44 | 21771 | 94 | ||
| NM_001306524.2 | NP_001293453.1 | 11 - 12 | 3722 | 94 | ||
| NM_001306525.3 | NP_001293454.1 | 11 - 12 | 3771 | 94 | ||
| NM_001356807.3 | NP_001343717.1 | 44 | 21531 | 94 | ||
| NM_001356808.3 | NP_001343716.1 | 45 | 21972 | 94 | ||
| NM_001356809.3 | NP_001343714.1 | 44 | 23625 | 94 | ||
| NM_001356812.3 | NP_001343617.1 | 36 - 37 | 22272 | 94 | ||
| NM_001356813.3 | NP_001343719.1 | 35 - 36 | 20112 | 94 | ||
| NM_001356814.3 | NP_001343711.1 | 36 - 37 | 20553 | 94 | ||
| NM_001356815.3 | NP_001343712.1 | 35 - 36 | 22206 | 94 | ||
| NM_001356816.3 | NP_001343713.1 | 34 - 35 | 20352 | 94 | ||
More Information
Set Membership: |
|

unc-89
Genomic Map