Assay Details
| Gene Symbol: unc-89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025814.4 | NP_001020985.1 | 40 - 41 | 21517 | 67 | |
| NM_001025818.3 | NP_001020989.1 | 38 - 39 | 19597 | 67 | ||
| NM_001306524.2 | NP_001293453.1 | 6 - 7 | 1548 | 67 | ||
| NM_001306525.3 | NP_001293454.1 | 6 - 7 | 1597 | 67 | ||
| NM_001356807.3 | NP_001343717.1 | 39 | 19357 | 67 | ||
| NM_001356808.3 | NP_001343716.1 | 40 | 19798 | 67 | ||
| NM_001356809.3 | NP_001343714.1 | 39 | 21451 | 67 | ||
| NM_001356812.3 | NP_001343617.1 | 31 - 32 | 20098 | 67 | ||
| NM_001356813.3 | NP_001343719.1 | 30 - 31 | 17938 | 67 | ||
| NM_001356814.3 | NP_001343711.1 | 31 - 32 | 18379 | 67 | ||
| NM_001356815.3 | NP_001343712.1 | 30 - 31 | 20032 | 67 | ||
| NM_001356816.3 | NP_001343713.1 | 29 - 30 | 18178 | 67 | ||
More Information
Set Membership: |
|

unc-89
Genomic Map