Assay Details
| Gene Symbol: hop-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058927.2 | NP_491328.1 | 1 - 2 | 137 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hop-1| Gene Symbol: hop-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058927.2 | NP_491328.1 | 1 - 2 | 137 | 70 | |
Set Membership: |
|