Assay Details
| Gene Symbol: nab-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379751.2 | NP_001367535.1 | 5 - 6 | 583 | 62 | |
| NM_001382891.1 | 3 - 4 | 774 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nab-1| Gene Symbol: nab-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379751.2 | NP_001367535.1 | 5 - 6 | 583 | 62 | |
| NM_001382891.1 | 3 - 4 | 774 | 62 | |||
Set Membership: |
|