Assay Details
| Gene Symbol: nab-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379751.2 | NP_001367535.1 | 7 - 8 | 956 | 97 | |
| NM_001379754.2 | NP_001367538.1 | 6 - 7 | 765 | 97 | ||
| NM_001382888.1 | 4 - 5 | 481 | 97 | |||
| NM_001382891.1 | 5 - 6 | 1147 | 97 | |||
| NM_001382892.1 | 7 - 8 | 957 | 97 | |||
More Information
Set Membership: |
|

nab-1
Genomic Map