Assay Details
| Gene Symbol: nab-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379751.2 | NP_001367535.1 | 10 - 11 | 2144 | 144 | |
| NM_001379754.2 | NP_001367538.1 | 9 - 10 | 1953 | 144 | ||
| NM_001382888.1 | 7 - 8 | 1669 | 144 | |||
| NM_001382891.1 | 8 - 9 | 2335 | 144 | |||
| NM_001382892.1 | 10 - 11 | 2145 | 144 | |||
More Information
Set Membership: |
|

nab-1
Genomic Map