Assay Details
| Gene Symbol: exoc-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058936.5 | NP_491337.1 | 6 - 7 | 1656 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

exoc-7| Gene Symbol: exoc-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058936.5 | NP_491337.1 | 6 - 7 | 1656 | 80 | |
Set Membership: |
|