Assay Details
| Gene Symbol: tufm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058937.6 | NP_491338.2 | 1 - 2 | 229 | 118 | |
| GenBank mRNA | AB010028.1 | - | 1 - 2 | 236 | 118 | |
More Information
Set Membership: |
|

tufm-2
Genomic Map