Assay Details
| Gene Symbol: met-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058939.6 | NP_491340.2 | 10 - 11 | 3253 | 85 | |
| NM_182042.4 | NP_871842.2 | 10 - 11 | 3224 | 85 | ||
More Information
Set Membership: |
|

met-1
Genomic Map