Assay Details
| Gene Symbol: met-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058939.6 | NP_491340.2 | 12 - 13 | 4142 | 95 | |
| NM_182042.4 | NP_871842.2 | 12 - 13 | 4113 | 95 | ||
More Information
Set Membership: |
|

met-1
Genomic Map