Assay Details
| Gene Symbol: met-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058939.6 | NP_491340.2 | 13 - 14 | 4937 | 121 | |
| NM_182042.4 | NP_871842.2 | 13 - 14 | 4908 | 121 | ||
More Information
Set Membership: |
|

met-1
Genomic Map