Assay Details
| Gene Symbol: met-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058939.6 | NP_491340.2 | 3 - 4 | 466 | 83 | |
| NM_182042.4 | NP_871842.2 | 3 - 4 | 437 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

met-1| Gene Symbol: met-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058939.6 | NP_491340.2 | 3 - 4 | 466 | 83 | |
| NM_182042.4 | NP_871842.2 | 3 - 4 | 437 | 83 | ||
Set Membership: |
|