Assay Details
| Gene Symbol: met-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058939.6 | NP_491340.2 | 9 - 10 | 2892 | 73 | |
| NM_182042.4 | NP_871842.2 | 9 - 10 | 2863 | 73 | ||
More Information
Set Membership: |
|

met-1
Genomic Map