Assay Details
| Gene Symbol: mus-81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382899.1 | 2 - 3 | 196 | 99 | ||
| NM_058940.7 | NP_491341.1 | 2 - 3 | 194 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382899.1 transcripts.
Set Membership: |
|

mus-81
Genomic Map