Assay Details
| Gene Symbol: tiam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058966.7 | NP_491367.3 | 2 - 3 | 202 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tiam-1| Gene Symbol: tiam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058966.7 | NP_491367.3 | 2 - 3 | 202 | 71 | |
Set Membership: |
|