Assay Details
| Gene Symbol: nrd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059002.6 | NP_491403.1 | 8 - 9 | 2813 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nrd-1| Gene Symbol: nrd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059002.6 | NP_491403.1 | 8 - 9 | 2813 | 92 | |
Set Membership: |
|