Assay Details
| Gene Symbol: got-2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059012.6 | NP_491413.2 | 1 - 2 | 196 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

got-2.1| Gene Symbol: got-2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059012.6 | NP_491413.2 | 1 - 2 | 196 | 121 | |
Set Membership: |
|