Assay Details
| Gene Symbol: lmd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059014.3 | NP_491415.2 | 1 - 2 | 121 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lmd-2| Gene Symbol: lmd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059014.3 | NP_491415.2 | 1 - 2 | 121 | 64 | |
Set Membership: |
|