Assay Details
| Gene Symbol: pdxk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001262804.4 | NP_001249733.1 | 1 - 2 | 129 | 79 | |
| NM_059062.3 | NP_491463.2 | 1 - 2 | 116 | 79 | ||
| NM_059063.6 | NP_491464.2 | 2 - 3 | 176 | 79 | ||
More Information
Set Membership: |
|

pdxk-1
Genomic Map