Assay Details
| Gene Symbol: pdxk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001262804.4 | NP_001249733.1 | 4 - 5 | 418 | 63 | |
| NM_059062.3 | NP_491463.2 | 4 - 5 | 435 | 63 | ||
| NM_059063.6 | NP_491464.2 | 5 - 6 | 465 | 63 | ||
More Information
Set Membership: |
|

pdxk-1
Genomic Map