Assay Details
| Gene Symbol: paf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383031.1 | 9 - 10 | 2304 | 113 | ||
| GenBank mRNA | AF386744.1 | - | 4 - 5 | 458 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

paf-1| Gene Symbol: paf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383031.1 | 9 - 10 | 2304 | 113 | ||
| GenBank mRNA | AF386744.1 | - | 4 - 5 | 458 | 113 | |
Set Membership: |
|