Assay Details
| Gene Symbol: cil-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059092.7 | NP_491493.2 | 2 - 3 | 290 | 82 | |
| Gene Symbol: paf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383031.1 | 1 - 2 | 129 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383031.1 transcripts.
Set Membership: |
|

cil-7
Genomic Map